Learn More
Medchemexpress LLC BAFF/TNFSF13B Trimer 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77571100UG
Encompass_Preferred
BAFF/TNFSF13B protein is a member of the TNF family and regulates immune responses and cell survival It is also known as B cell activating factor which plays a vital role in the development activation and survival of B cells by binding to receptors such as BAFF-R BAFF/TNFSF13B Trimer Protein Cynomolgus (HEK293 His-Avi) is the recombinant cynomolgus-derived BAFF/TNFSF13B protein expressed by HEK293 with N-His N-Avi labeled tag
Catalog No. 50-001-87126