Learn More
Medchemexpress LLC BAFF/TNFSF13B Trimer 10ug | 10ug
SDP
Catalog No. 5000187227
Encompass_Preferred
BAFF/TNFSF13B protein is a member of the TNF family and regulates immune responses and cell survival It is also known as B cell activating factor which plays a vital role in the development activation and survival of B cells by binding to receptors such as BAFF-R BAFF/TNFSF13B Trimer Protein Cynomolgus (HEK293 His-Avi) is the recombinant cynomolgus-derived BAFF/TNFSF13B protein expressed by HEK293 with N-His N-Avi labeled tag
Catalog No. 50-001-87227 Supplier Medchemexpress LLC Supplier No. HYP7757110UG
Add to Cart
Add to Cart
BAFF/TNFSF13B protein is a member of the TNF family and regulates immune responses and cell survival It is also known as B cell activating factor which plays a vital role in the development activation and survival of B cells by binding to receptors such as BAFF-R BAFF/TNFSF13B Trimer Protein Cynomolgus (HEK293 His-Avi) is the recombinant cynomolgus-derived BAFF/TNFSF13B protein expressed by HEK293 with N-His N-Avi labeled tag