Learn More
Medchemexpress LLC BAFF/TNFSF13B Trimer 20ug | 20ug
SDP
Catalog No. 5000187048
Encompass_Preferred
BAFF/TNFSF13B protein is a member of the TNF family and regulates immune responses and cell survival It is also known as B cell activating factor which plays a vital role in the development activation and survival of B cells by binding to receptors such as BAFF-R BAFF/TNFSF13B Trimer Protein Cynomolgus (HEK293 His-Avi) is the recombinant cynomolgus-derived BAFF/TNFSF13B protein expressed by HEK293 with N-His N-Avi labeled tag
Catalog No. 50-001-87048 Supplier Medchemexpress LLC Supplier No. HYP7757120UG
BAFF/TNFSF13B protein is a member of the TNF family and regulates immune responses and cell survival It is also known as B cell activating factor which plays a vital role in the development activation and survival of B cells by binding to receptors such as BAFF-R BAFF/TNFSF13B Trimer Protein Cynomolgus (HEK293 His-Avi) is the recombinant cynomolgus-derived BAFF/TNFSF13B protein expressed by HEK293 with N-His N-Avi labeled tag