Learn More
Medchemexpress LLC BAFF/TNFSF13B Trimer 50ug | 50ug
SDP
Catalog No. 5000194017
Encompass_Preferred
BAFF/TNFSF13B protein is a cytokine that binds to TNFRSF13B/TACI and TNFRSF17/BCMA forming a key ligand-receptor pathway together with TNFSF13/APRIL These interactions play a crucial role in stimulating B-cell and T-cell function and regulating humoral immunity BAFF/TNFSF13B Trimer Protein Human (HEK293 His-Flag) is the recombinant human-derived BAFF/TNFSF13B protein expressed by HEK293 with N-His N-Flag labeled tag
Catalog No. 50-001-94017 Supplier Medchemexpress LLC Supplier No. HYP7838450UG
BAFF/TNFSF13B protein is a cytokine that binds to TNFRSF13B/TACI and TNFRSF17/BCMA forming a key ligand-receptor pathway together with TNFSF13/APRIL These interactions play a crucial role in stimulating B-cell and T-cell function and regulating humoral immunity BAFF/TNFSF13B Trimer Protein Human (HEK293 His-Flag) is the recombinant human-derived BAFF/TNFSF13B protein expressed by HEK293 with N-His N-Flag labeled tag