Learn More
Bellco Glass, Inc. Baffle Assembly,15L For 10mm Port
SDP
Catalog No. 501946481
Encompass_Preferred
Size: 15lFor 10mm portMaterial: 316l sst with ra finish of 32? Or betterDimensions: 3/8" od shaft with 1" wide baffleInsertion length: 15 ½". MADE IN THE USA
Catalog No. 50-194-6481 Supplier Bellco Glass, Inc. Supplier No. 790915077
Size: 15lFor 10mm portMaterial: 316l sst with ra finish of 32? Or betterDimensions: 3/8" od shaft with 1" wide baffleInsertion length: 15 ½". MADE IN THE USA