Learn More
Chemglass Life Sciences Complete Baffle System: Overall Baffle Length: 515mm, Effective Baffle Length: 315mm, Baffle Width x Depth (mm): 7 x 10, PTFE Compression Fitting: 1/4", Adapter Joint Size: 24/40
SDP
Catalog No. CG1971M04
Encompass_Preferred
Complete Baffle System: Overall Baffle Length: 515mm, Effective Baffle Length: 315mm, Baffle Width x Depth (mm): 7 x 10, PTFE Compression Fitting: 1/4", Adapter Joint Size: 24/40. Unique baffle system for benchtop process reactors from 300mL to 5L. Designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors. PTFE jacketed stainless steel provides excellent strength and chemical durability.Features:¿ Adjustable Rectangular Baffle¿ PTFE Standard Taper Adapter Is Offset Permitting Baffle To Be Adjusted Up To 1/4" Towards The Center Of The Reactor.¿ Baffle Can Be Vertically Adjusted Inside Reactor.Complete Baffles Consist of: PTFE Jacketed S.S. Baffle, PTFE Standard Taper Adapter, And A 1/4" PTFE Compression Fitting.The 360mm Overall Baffle Length Is For 300mL-500mL Reactors.The 410mm Overall Baffle Length Is For 1L, 2L, & 4L Reactors.The 515mm Overall Baffle Length Is For 3L & 5L Reactors.
Catalog No. CG1971M04 Supplier Chemglass Life Sciences Supplier No. CG1971M04
Complete Baffle System: Overall Baffle Length: 515mm, Effective Baffle Length: 315mm, Baffle Width x Depth (mm): 7 x 10, PTFE Compression Fitting: 1/4", Adapter Joint Size: 24/40. Unique baffle system for benchtop process reactors from 300mL to 5L. Designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors. PTFE jacketed stainless steel provides excellent strength and chemical durability.Features:¿ Adjustable Rectangular Baffle¿ PTFE Standard Taper Adapter Is Offset Permitting Baffle To Be Adjusted Up To 1/4" Towards The Center Of The Reactor.¿ Baffle Can Be Vertically Adjusted Inside Reactor.Complete Baffles Consist of: PTFE Jacketed S.S. Baffle, PTFE Standard Taper Adapter, And A 1/4" PTFE Compression Fitting.The 360mm Overall Baffle Length Is For 300mL-500mL Reactors.The 410mm Overall Baffle Length Is For 1L, 2L, & 4L Reactors.The 515mm Overall Baffle Length Is For 3L & 5L Reactors.