Learn More
Bellco Glass, Inc. Baffle Assembly, 3L For 6mm Port
SDP
Catalog No. 501946433
Encompass_Preferred
Baffle Assembly 3L. For 6mm Port. Material: 316L SST with electropolish finish - RA 32 - outside. Dimensions of baffle: 5/8" OD x 7" long. Insertion of baffle assy: 9 1/8". Supply with: A555-7 . MADE IN THE USA
Catalog No. 50-194-6433 Supplier Bellco Glass, Inc. Supplier No. 790900022
Baffle Assembly 3L. For 6mm Port. Material: 316L SST with electropolish finish - RA 32 - outside. Dimensions of baffle: 5/8" OD x 7" long. Insertion of baffle assy: 9 1/8". Supply with: A555-7 . MADE IN THE USA