Learn More
Bellco Glass, Inc. Baffle Assembly,7L For 10mm Port
SDP
Catalog No. 501946446
Encompass_Preferred
316L stainless steel baffle assembly for use on large flange bioreactor vesselsystem. Attaches to the head plate. Used to change the mixing characteristics within the bioreactor.--Size: 7L. MADE IN THE USA
Catalog No. 50-194-6446 Supplier Bellco Glass, Inc. Supplier No. 790900039
316L stainless steel baffle assembly for use on large flange bioreactor vesselsystem. Attaches to the head plate. Used to change the mixing characteristics within the bioreactor.--Size: 7L. MADE IN THE USA