Learn More
Chemglass Life Sciences Complete Baffles for use in 10L-20L Reactors: Overall Baffle Length: 32", Effective Baffle Length: 13.5", Baffle Width x Depth (mm): 20 x 30, Closed Bottom, Adapter Joint Size: 45/50
SDP
Catalog No. CG1971120
Encompass_Preferred
Complete Baffles for use in 10L-20L Reactors: Overall Baffle Length: 32", Effective Baffle Length: 13.5", Baffle Width x Depth (mm): 20 x 30, Lower PTFE Compression Fitting: Closed Bottom, Adapter Joint Size: 45/50. Unique glass baffle system for process reactors from 2L to 50L. Designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors. All baffles have 3/4" precision ground top and lower tubing for use with compression fittings and top jointed adapter. PLEASE NOTE: SEE CG-1872-B FOR pH ELECTRODE BAFFLES.Features:¿Available In Square Or Rectangular Shapes, Open Or Closed Bottom¿Baffle Can Be Vertically Adjusted Inside Reactor¿Hollow Baffles Allow Thermocouples From 1/8" to 1/4" In Diameter To Be Inserted Into The ReactionComplete Baffles Consist Of: Glass Baffle, Glass Fitted PTFE Adapter, And Lower PTFE Compression Fitting.¿Designed and developed by John Korzun, Bristol Myers Squibb, New Brunswick, NJ
Catalog No. CG1971120 Supplier Chemglass Life Sciences Supplier No. CG1971120
Complete Baffles for use in 10L-20L Reactors: Overall Baffle Length: 32", Effective Baffle Length: 13.5", Baffle Width x Depth (mm): 20 x 30, Lower PTFE Compression Fitting: Closed Bottom, Adapter Joint Size: 45/50. Unique glass baffle system for process reactors from 2L to 50L. Designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors. All baffles have 3/4" precision ground top and lower tubing for use with compression fittings and top jointed adapter. PLEASE NOTE: SEE CG-1872-B FOR pH ELECTRODE BAFFLES.Features:¿Available In Square Or Rectangular Shapes, Open Or Closed Bottom¿Baffle Can Be Vertically Adjusted Inside Reactor¿Hollow Baffles Allow Thermocouples From 1/8" to 1/4" In Diameter To Be Inserted Into The ReactionComplete Baffles Consist Of: Glass Baffle, Glass Fitted PTFE Adapter, And Lower PTFE Compression Fitting.¿Designed and developed by John Korzun, Bristol Myers Squibb, New Brunswick, NJ