Learn More
Bellco Glass, Inc. Baffle Assembly , 7L Large Flange
SDP
Catalog No. 501944807
Encompass_Preferred
316l stainless steel baffle assembly for use on large flange 7 liter bioreactor vessel system. Attaches to the head plate. used to change the mixing characteristics within the bioreactor. MADE IN THE USA. 3 required per vessel 1/4" rod Includes: Silicone 0-ring
Catalog No. 50-194-4807 Supplier Bellco Glass, Inc. Supplier No. 196453600
316l stainless steel baffle assembly for use on large flange 7 liter bioreactor vessel system. Attaches to the head plate. used to change the mixing characteristics within the bioreactor. MADE IN THE USA. 3 required per vessel 1/4" rod Includes: Silicone 0-ring