Learn More
Ace Glass, Inc. Baffle, PTFE, fits #11 Ace-Thred, 330mm long, 18x150x10 baffle, can take 1/4in OD temp probe up to 500mm length.
SDP
Catalog No. 50977561
Encompass
18x150x10 PTFE baffle with an integral stainless steel tube. Can be used for either liquid addition into a reactor, or primarily as a temperature probe. Baffle fits a #11 Ace-Thred and is 330mm long. Baffle can take a 1/4 O.D. temperature probe that is up to 500mm long. The #7 Ace-Thred bottom bushing will make a tight compression seal with either the tubing or a thermocouple.
Catalog No. 50-977-561 Supplier Ace Glass, Inc. Supplier No. 1219303
18x150x10 PTFE baffle with an integral stainless steel tube. Can be used for either liquid addition into a reactor, or primarily as a temperature probe. Baffle fits a #11 Ace-Thred and is 330mm long. Baffle can take a 1/4 O.D. temperature probe that is up to 500mm long. The #7 Ace-Thred bottom bushing will make a tight compression seal with either the tubing or a thermocouple.