Learn More
Ace Glass, Inc. Baffle, PTFE, fits #25 Ace-Thred, 610mm long, 32x360x14 baffle, can take 1/4in OD temp probe up to 900mm length.
SDP
Catalog No. 50977562
Encompass
32x360x14 PTFE baffle with an integral stainless steel tube. Can be used for either liquid addition into a reactor, or primarily as a temperature probe. Baffle fits a #25 Ace-Thred and is 610mm long. Baffle can take a 1/4 O.D. temperature probe that is up to 900mm long. The #7 Ace-Thred bottom bushing will make a tight compression seal with either the tubing or a thermocouple.
Catalog No. 50-977-562 Supplier Ace Glass, Inc. Supplier No. 1219309
32x360x14 PTFE baffle with an integral stainless steel tube. Can be used for either liquid addition into a reactor, or primarily as a temperature probe. Baffle fits a #25 Ace-Thred and is 610mm long. Baffle can take a 1/4 O.D. temperature probe that is up to 900mm long. The #7 Ace-Thred bottom bushing will make a tight compression seal with either the tubing or a thermocouple.