Learn More
Chemglass Life Sciences PTFE Jacketed Stainless Steel Baffles Only: Overall Baffle Length: 410mm, Effective Baffle Length: 220mm, Baffle Width x Depth (mm): 7 x 10, PTFE Compression Fitting: 1/4", Fits Reactor Size: 1L, 2L, & 4L
SDP
Catalog No. 501531356
Encompass_Preferred
PTFE Jacketed Stainless Steel Baffles Only: Overall Baffle Length: 410mm, Effective Baffle Length: 220mm, Baffle Width x Depth (mm): 7 x 10, PTFE Compression Fitting: 1/4", Fits Reactor Size: 1L, 2L, & 4L. Designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors. PTFE jacketed Stainless Steel provides excellent strength and chemical durability. Pictured is a fully assembled baffle system, but this item is BAFFLE ONLY.
Catalog No. 50-153-1356 Supplier Chemglass Life Sciences Supplier No. CG1971M10
PTFE Jacketed Stainless Steel Baffles Only: Overall Baffle Length: 410mm, Effective Baffle Length: 220mm, Baffle Width x Depth (mm): 7 x 10, PTFE Compression Fitting: 1/4", Fits Reactor Size: 1L, 2L, & 4L. Designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors. PTFE jacketed Stainless Steel provides excellent strength and chemical durability. Pictured is a fully assembled baffle system, but this item is BAFFLE ONLY.