Learn More
Alkali Scientific Baffled 250mL Erlenmeyer Flask with Vented Cap and Flat Bottom, PETG bottle, Sterile, 12/cs
SDP
Catalog No. 502335943
Encompass_Preferred
Baffled interior bottoms provide a swirl diruption/break which improves the aeration of the culture. This feature can be beneficial to use when the cultures' oxygen requirements are high or viscous. Baffled interior bottoms provide a swirl diruption/break which improves the aeration of the culture. This feature can be beneficial to use when the cultures' oxygen requirements are high or viscous.
Catalog No. 50-233-5943 Supplier Alkali Scientific Supplier No. EFB304
Baffled interior bottoms provide a swirl diruption/break which improves the aeration of the culture. This feature can be beneficial to use when the cultures' oxygen requirements are high or viscous. Baffled interior bottoms provide a swirl diruption/break which improves the aeration of the culture. This feature can be beneficial to use when the cultures' oxygen requirements are high or viscous.