Learn More
Alkali Scientific Baffled 500mL Erlenmeyer Flask with Vented Cap and Flat Bottom, PETG, Sterile, 12/cs
SDP
Catalog No. 502336055
Encompass_Preferred
Baffled interior bottoms provide a swirl diruption/break which improves the aeration of the culture. This feature can be beneficial to use when the cultures' oxygen requirements are high or viscous.
Catalog No. 50-233-6055 Supplier Alkali Scientific Supplier No. EFB305
Baffled interior bottoms provide a swirl diruption/break which improves the aeration of the culture. This feature can be beneficial to use when the cultures' oxygen requirements are high or viscous.