Learn More
Perkin Elmer US LLC Baffled Quartz Cyclonic Spray Chamber with PFA Torch Adapter for Avio 200
Catalog No. 502113152
Encompass_Preferred
The Baffled Cyclonic Spray Chamber features a central transfer tube which acts as a secondary particle separator, helping to separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits.
Catalog No. 50-211-3152 Supplier Perkin Elmer US LLC Supplier No. N0791842
The Baffled Cyclonic Spray Chamber features a central transfer tube which acts as a secondary particle separator, helping to separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits.