Learn More
ACROBiosystems Recombinant Protein;Mouse BAFF / TNFSF13B / CD257 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;BAF,TNFSF13B,BAFF,BLYS,CD257,DTL,TALL-1,TALL1,THANK,TNFSF20,ZTNF4,TNFSF-13B,ZTNF-4
Supplier:  ACROBiosystems BAFM5257100UG
Encompass_Preferred
Recombinant Protein;Mouse BAFF / TNFSF13B / CD257 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Mouse BAFF, Mouse IgG2a Fc Tag, low endotoxin (BAF-M5257) is expressed from human 293 cells (HEK293). It contains AA Ala 127 - Leu 309 (Accession # Q9WU72-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 47.7 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse BAFF, Mouse IgG2a Fc Tag (Cat. No. BAF-M5257) at 5 μg/mL (100 μL/well) can bind Mouse BCMA, Fc Tag (Cat. No. BCA-M5258) with a linear range of 2-10 ng/mL (QC tested).
Catalog No. 50-165-8100