Learn More
ACROBiosystems Recombinant Protein;Mouse BAFFR / TNFRSF13C Protein, Fc Tag;HEK293;1MG;BAR,BAFFR,TNFRSF13C,BAFF-R,BROMIX,CD268,CVID4,prolixin
Catalog No. 501658431
Encompass_Preferred
Recombinant Protein;Mouse BAFFR / TNFRSF13C Protein, Fc Tag;1MG;Mouse BAFFR, Fc Tag (BAR-M5259) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Ala 71 (Accession # NP_082351.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 33.3 kDa. As a result of glycosylation, the protein migrates as 40-45 kDa under reducing (R) condition, and 80-100 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 5 μg/mL (100 μL/well) can bind Mouse BAFFR, Fc Tag (Cat. No. BAR-M5259) with a linear range of 0.3-13 ng/mL (QC tested).
Catalog No. 50-165-8431 Supplier ACROBiosystems Supplier No. BARM52591MG
Recombinant Protein;Mouse BAFFR / TNFRSF13C Protein, Fc Tag;1MG;Mouse BAFFR, Fc Tag (BAR-M5259) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Ala 71 (Accession # NP_082351.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 33.3 kDa. As a result of glycosylation, the protein migrates as 40-45 kDa under reducing (R) condition, and 80-100 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 5 μg/mL (100 μL/well) can bind Mouse BAFFR, Fc Tag (Cat. No. BAR-M5259) with a linear range of 0.3-13 ng/mL (QC tested).