Learn More
ACROBiosystems Recombinant Protein;Human BAFFR / TNFRSF13C Protein, Fc Tag;HEK293;1MG;BAR,BAFFR,TNFRSF13C,BAFF-R,BROMIX,CD268,CVID4,prolixin
Catalog No. 501658408
Encompass_Preferred
Recombinant Protein;Human BAFFR / TNFRSF13C Protein, Fc Tag;1MG;Human BAFFR, Fc Tag (BAR-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 33.2 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 5 μg/mL (100 μL/well) can bind Human BAFFR, Fc Tag (Cat. No. BAR-H5257) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-165-8408 Supplier ACROBiosystems Supplier No. BARH52571MG
Recombinant Protein;Human BAFFR / TNFRSF13C Protein, Fc Tag;1MG;Human BAFFR, Fc Tag (BAR-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession # Q96RJ3-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 33.2 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 5 μg/mL (100 μL/well) can bind Human BAFFR, Fc Tag (Cat. No. BAR-H5257) with a linear range of 2-31 ng/mL (QC tested).