Learn More
Lkt Laboratories CAS 88899-55-2, ≥90%, Vacuolar H+ ATPase inhibitor. It suppresses autophagy, prevents tumor growth in breast cancer models, and inhibits growth of Plasmodium.
SDP
Catalog No. NC2530058
Encompass_Preferred
Bafilomycin A1 is a macrolide antibiotic initially produced by Streptomyces that exhibits anti-parasitic, antimalarial, and anticancer chemotherapeutic activities. Bafilomycin A1 inhibits the vacuolar H+ ATPase and suppresses autophagy. Bafilomycin kills Plasmodium parasites in cellular models, and separately, induces cell death and suppresses tumor growth in animal models of breast cancer.
Catalog No. NC2530058 Supplier Lkt Laboratories Supplier No. B0025
Bafilomycin A1 is a macrolide antibiotic initially produced by Streptomyces that exhibits anti-parasitic, antimalarial, and anticancer chemotherapeutic activities. Bafilomycin A1 inhibits the vacuolar H+ ATPase and suppresses autophagy. Bafilomycin kills Plasmodium parasites in cellular models, and separately, induces cell death and suppresses tumor growth in animal models of breast cancer.