Learn More
Research Products International Corp Bafilomycin A1, 1 Milligram
SDP
Catalog No. 50995800
Encompass_Preferred
Bafilomycin A1 is a potent and selective inhibitor of the vacuolar-type H+-ATPase (V-ATPase), an enzyme responsible for maintaining acidic pH in cellular compartments such as lysosomes and endosomes.
In the life science field, Bafilomycin A1 is primarily used as a research tool to investigate the role of V-ATPase and lysosomal acidification in various cellular processes. By inhibiting V-ATPase, Bafilomycin A1 disrupts the acidification of cellular compartments, which in turn affects processes such as protein degradation, autophagy, endocytosis, and vesicular trafficking. 
Bafilomycin A1 can be used to study autophagy, a cellular process involved in the degradation and recycling of cellular components. By inhibiting V-ATPase, Bafilomycin A1 blocks the final step of autophagy, preventing the fusion of autophagosomes with lysosomes and thereby inhibiting autophagic degradation.
Catalog No. 50-995-800 Supplier Research Products International Corp Supplier No. B405000.001
Bafilomycin A1 is a potent and selective inhibitor of the vacuolar-type H+-ATPase (V-ATPase), an enzyme responsible for maintaining acidic pH in cellular compartments such as lysosomes and endosomes.
In the life science field, Bafilomycin A1 is primarily used as a research tool to investigate the role of V-ATPase and lysosomal acidification in various cellular processes. By inhibiting V-ATPase, Bafilomycin A1 disrupts the acidification of cellular compartments, which in turn affects processes such as protein degradation, autophagy, endocytosis, and vesicular trafficking. 
Bafilomycin A1 can be used to study autophagy, a cellular process involved in the degradation and recycling of cellular components. By inhibiting V-ATPase, Bafilomycin A1 blocks the final step of autophagy, preventing the fusion of autophagosomes with lysosomes and thereby inhibiting autophagic degradation.