Learn More
Medchemexpress LLC HY-100558 1mg Medchemexpress, Bafilomycin A1 CAS:88899-55-2 Purity:>98%
SDP
Catalog No. 501967747
Encompass_Preferred
Medchemexpress, HY-100558 1mg Bafilomycin A1 CAS:88899-55-2 Bafilomycin A1, a macrolide antibiotic isolated from the Streptomyces species, is a specific inhibitor of vacuolar-type H+ ATPase (V-ATPase). Bafilomycin A1 inhibits autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7747 Supplier Medchemexpress LLC Supplier No. HY1005581MG
Medchemexpress, HY-100558 1mg Bafilomycin A1 CAS:88899-55-2 Bafilomycin A1, a macrolide antibiotic isolated from the Streptomyces species, is a specific inhibitor of vacuolar-type H+ ATPase (V-ATPase). Bafilomycin A1 inhibits autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.