Learn More
TARGETMOL CHEMICALS INC BAFILOMYCIN A1 5MG
SDP
Catalog No. 502579296
Encompass_Preferred
Also available in 1 mg 160 uL * 1 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bafilomycin A1 induces caspase-independent cell death in hepatocellular carcinoma cells via targeting of autophagy and MAPK pathways. Bafilomycin A1 is a vacuolar H+-ATPase inhibitor (IC50 0.44 nM). It is also found to inhibit autophagy while induces apoptosis.Bafilomycin A1 triggers proliferative potential of senescent cancer cells in vitro and in NOD/SCID mice. purity: 98%
Catalog No. 50-257-9296 Supplier TARGETMOL CHEMICALS INC Supplier No. T67405MG
Also available in 1 mg 160 uL * 1 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bafilomycin A1 induces caspase-independent cell death in hepatocellular carcinoma cells via targeting of autophagy and MAPK pathways. Bafilomycin A1 is a vacuolar H+-ATPase inhibitor (IC50 0.44 nM). It is also found to inhibit autophagy while induces apoptosis.Bafilomycin A1 triggers proliferative potential of senescent cancer cells in vitro and in NOD/SCID mice. purity: 98%