Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bafilomycin A1 | 88899-55-2 | MFCD06795130 | 100.0% | 622.83 g/mol | C35H58O9 | 10 MG
SDP

Catalog No. 5000212729
Encompass_Preferred

Bafilomycin A1 is a macrolide antibiotic that specifically and reversibly inhibits vacuolar H+-ATPase (V-ATPase). It is widely used in cell biology to inhibit lysosomal acidification and to block autophagosome-lysosome fusion, making it a standard reagent for studies of autophagy, apoptosis, and membrane transport processes.

  • Specific, reversible inhibitor of vacuolar H+-ATPase.
  • Blocks autophagosome-lysosome fusion and inhibits lysosomal acidification.
  • Commonly used in autophagy, apoptosis, and membrane transporter studies.
  • High solubility in DMSO (≥100 mg/mL); in vivo formulation protocols provided.
  • Recommended storage -20°C, protect from light; in solvent -80°C for up to 6 months.
  • Molecular weight 622.83 g/mol; CAS 88899-55-2.

Catalog No. 50-002-12729 Supplier Medchemexpress LLC Supplier No. HY10055810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bafilomycin A1 is a macrolide antibiotic that specifically and reversibly inhibits vacuolar H+-ATPase (V-ATPase). It is widely used in cell biology to inhibit lysosomal acidification and to block autophagosome-lysosome fusion, making it a standard reagent for studies of autophagy, apoptosis, and membrane transport processes.

  • Specific, reversible inhibitor of vacuolar H+-ATPase.
  • Blocks autophagosome-lysosome fusion and inhibits lysosomal acidification.
  • Commonly used in autophagy, apoptosis, and membrane transporter studies.
  • High solubility in DMSO (≥100 mg/mL); in vivo formulation protocols provided.
  • Recommended storage -20°C, protect from light; in solvent -80°C for up to 6 months.
  • Molecular weight 622.83 g/mol; CAS 88899-55-2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.