Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bafilomycin A1, 95%

Catalog No. ac328120001
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328120001 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC328120001 Supplier Thermo Scientific Chemicals Supplier No. 328120001
Only null left
Add to Cart
This item is not returnable. View return policy

Bafilomycin A1, CAS # 88899-55-2, is an inhibitor of vacuolar ATPases from Streptomyces griseus that has antibacterial, antifungal, and antineoplastic activity. | CAS: 88899-55-2 | C35H58O9 | 622.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Bafilomycin A1 is a specific potent inhibitor of vacuolar ATPases
  • It is a macrolide antibiotic, and shows fungicide activity

Application

  • Bafilomycin A1 has antibacterial, antifungal, and antineoplastic activity
  • It can be immunosuppressive
  • It can prevent maturation of autophagic vacuoles by inhibiting fusion between autophagosomes and lysosomes
  • It is a potent and selective inhibitor of vacuolar-type H+ ATPase (V-ATPase)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88899-55-2
Molecular Formula C35H58O9
Molecular Weight (g/mol) 622.84
MDL Number MFCD06795130
InChI Key XDHNQDDQEHDUTM-XRCNURIOSA-N
PubChem CID 133268469
IUPAC Name (3Z,5E,7S,8R,9R,11Z,15R,16S)-16-[(2R,3S,4R)-4-[(2S,4S,5R,6S)-2,4-dihydroxy-5-methyl-6-propan-2-yloxan-2-yl]-3-hydroxypentan-2-yl]-8-hydroxy-3,15-dimethoxy-5,7,9,11-tetramethyl-1-oxacyclohexadeca-3,5,11,13-tetraen-2-one
SMILES CC1CC(=CC=CC(C(OC(=O)C(=CC(=CC(C1O)C)C)OC)C(C)C(C(C)C2(CC(C(C(O2)C(C)C)C)O)O)O)OC)C

Specifications

Type Bafilomycin A1
Physical Form Powder/Solid
Color White
Quantity 0.1 mg
Assay Percent Range 95%
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in methanol, ethanol, dmso
Formula Weight 622.8
Percent Purity 95%
Chemical Name or Material Bafilomycin A1, 95%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.