Learn More
Selleck Chemical LLC Bafilomycin A1 (Baf-A1) 1mg 88899-55-2
SDP
Catalog No. 501361229
Encompass_Preferred
Bafilomycin A1(Baf-A1) is a vacuolar H+-ATPase inhibitor with IC50 of 0.44 nM. Bafilomycin A1 is found to inhibit autophagy while induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1229 Supplier Selleck Chemical LLC Supplier No. S14131MG
Bafilomycin A1(Baf-A1) is a vacuolar H+-ATPase inhibitor with IC50 of 0.44 nM. Bafilomycin A1 is found to inhibit autophagy while induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.