Learn More
Enzo Life Sciences Bafilomycin B1 (1mg). CAS: 88899-56-3; 80112-34-1
SDP
Catalog No. 502008939
Encompass_Preferred
Macrolide antibiotic that acts as a specific inhibitor of vacuolar-type H+-ATPase. Purity: ≥95% (HPLC). Solubility: Slightly soluble in methanol or DMSO; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8939 Supplier Enzo Life Sciences Supplier No. ALX380063M001
Macrolide antibiotic that acts as a specific inhibitor of vacuolar-type H+-ATPase. Purity: ≥95% (HPLC). Solubility: Slightly soluble in methanol or DMSO; insoluble in water. Long Term Storage: -20°C.