Learn More
AdipoGen Bafilomycin B1
SDP
Catalog No. 502562157
Encompass_Preferred
Chemical. CAS 88899-56-3. Formula C44H65NO13. MW 815.9. Isolated from Streptomyces hygroscopicus. Macrolide antibiotic. Specific vacuolar-type H-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Antitrypanosomal and antileishmanial compound. Phytotoxic. Influences the cell wall biosynthesis of Asp. niger.
Catalog No. 50-256-2157 Supplier AdipoGen Supplier No. BVT0004C100
Chemical. CAS 88899-56-3. Formula C44H65NO13. MW 815.9. Isolated from Streptomyces hygroscopicus. Macrolide antibiotic. Specific vacuolar-type H-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Antitrypanosomal and antileishmanial compound. Phytotoxic. Influences the cell wall biosynthesis of Asp. niger.