Learn More
AdipoGen Bafilomycin C1
SDP
Supplier:  AdipoGen BVT0068M001
Encompass_Preferred
Chemical. CAS 88979-61-7. Formula C39H60O12. MW 720.9. Isolated from Streptomyces sp. Gö 14F. Antibiotic. Specific vacuolar-type H-ATPase inhibitor. Inhibitor of autophagic degradation by rising lysosomal pH and thus inactivating the lysosomal acid hydrolases. Antibacterial, antifungal, insecticidal and anthelmintic. Potential anti-osteoporotic agent in treating bone lytic diseases. Neuroprotection inducer.
Catalog No. 50-256-2120