Learn More
Medchemexpress LLC Bafilomycin C1 | 88979-61-7 | 99.9% | 1 MG
SDP
Catalog No. 5000340360
Encompass_Preferred
Bafilomycin C1 is a macrolide antibiotic isolated from Streptomyces sp. It is a potent, specific, and reversible inhibitor of vacuolar-type H+-ATPases (V-ATPases). Bafilomycin C1 inhibits the growth of gram-positive bacteria and fungi, induces cell apoptosis, and is used for studying hepatocellular carcinoma (HCC).
  • Potent, specific, and reversible inhibitor of V-ATPases
  • Isolated from Streptomyces sp.
  • Inhibits gram-positive bacteria and fungi
  • Induces cell apoptosis
  • Used for studying hepatocellular carcinoma (HCC)
  • Induces G0/G1 cell-cycle arrest
  • Induces mitochondrial-mediated apoptosis
  • 99.9% purity
Catalog No. 50-003-40360 Supplier Medchemexpress LLC Supplier No. HY1301731MG
Bafilomycin C1 is a macrolide antibiotic isolated from Streptomyces sp. It is a potent, specific, and reversible inhibitor of vacuolar-type H+-ATPases (V-ATPases). Bafilomycin C1 inhibits the growth of gram-positive bacteria and fungi, induces cell apoptosis, and is used for studying hepatocellular carcinoma (HCC).
  • Potent, specific, and reversible inhibitor of V-ATPases
  • Isolated from Streptomyces sp.
  • Inhibits gram-positive bacteria and fungi
  • Induces cell apoptosis
  • Used for studying hepatocellular carcinoma (HCC)
  • Induces G0/G1 cell-cycle arrest
  • Induces mitochondrial-mediated apoptosis
  • 99.9% purity