Learn More
Medchemexpress LLC Bafilomycin D | 98813-13-9 | 98.4% | 604.81 | 10 MG
SDP
Catalog No. 5000345755
Encompass_Preferred
Bafilomycin D is a specific inhibitor of vacuolar-type ATPase (V-ATPase). It exhibits antimicrobial, insecticidal, herbicidal, and cytotoxic activity.
  • Specific inhibitor of vacuolar-type ATPase (V-ATPase).
  • Antimicrobial activity.
  • Insecticidal activity.
  • Herbicidal activity.
  • Cytotoxic activity.
Catalog No. 50-003-45755 Supplier Medchemexpress LLC Supplier No. HY1089701MG
Bafilomycin D is a specific inhibitor of vacuolar-type ATPase (V-ATPase). It exhibits antimicrobial, insecticidal, herbicidal, and cytotoxic activity.
  • Specific inhibitor of vacuolar-type ATPase (V-ATPase).
  • Antimicrobial activity.
  • Insecticidal activity.
  • Herbicidal activity.
  • Cytotoxic activity.