Learn More
Medchemexpress LLC Bafilomycin D | 98813-13-9 | MFCD14635429 | 98.4% | 604.81 g/mol | C35H56O8 | 5 MG
SDP
Catalog No. 5000263496
Encompass_Preferred
Bafilomycin D is a macrolide natural product that acts as a specific inhibitor of vacuolar-type ATPase (V-ATPase). It is employed in biochemical and cell biology research to probe autophagy, endosomal and lysosomal acidification, and proton pump function, and it displays antimicrobial, insecticidal, herbicidal, and cytotoxic activities.
  • Specific inhibitor of vacuolar-type ATPase (V-ATPase).
  • Used to study autophagy and lysosomal function in cells.
  • Causes endosomal and lysosomal deacidification.
  • Provided as a high-purity solid suitable for research use.
  • Powder stable at -20°C; solutions recommended for -80°C storage for long term.
Catalog No. 50-002-63496 Supplier Medchemexpress LLC Supplier No. HY1089705MG
Bafilomycin D is a macrolide natural product that acts as a specific inhibitor of vacuolar-type ATPase (V-ATPase). It is employed in biochemical and cell biology research to probe autophagy, endosomal and lysosomal acidification, and proton pump function, and it displays antimicrobial, insecticidal, herbicidal, and cytotoxic activities.
  • Specific inhibitor of vacuolar-type ATPase (V-ATPase).
  • Used to study autophagy and lysosomal function in cells.
  • Causes endosomal and lysosomal deacidification.
  • Provided as a high-purity solid suitable for research use.
  • Powder stable at -20°C; solutions recommended for -80°C storage for long term.