Learn More
Andwin Scientific BAG 210L 36X45IN CLEAR 100CS
SDP
Catalog No. NC3779228
Encompass_Preferred
printed bag, without indicator, 210l, pp, 915x1150mm (36x45") 50um/2mil thickness,clear
Catalog No. NC3779228 Supplier Andwin Scientific Supplier No. 77778434
printed bag, without indicator, 210l, pp, 915x1150mm (36x45") 50um/2mil thickness,clear