Learn More
Uline BAG 7X8IN 6MIL RECLOS 1000/CS
Catalog No. NC3385353
Encompass_Preferred
7 x 8" 6 Mil Reclosable Bags - 1 Quart, 1000/cs Superior strength, puncture-resistant poly with heavy-duty zipper for security. 100% virgin polyethylene film. Meets FDA and USDA specifications. Latex free,BHT free (Butylated Hydroxy Toluene), Chlorine free, PVC free. TEMPERATURE: 20° - 150° F
Catalog No. NC3385353 Supplier Uline Supplier No. S15260
7 x 8" 6 Mil Reclosable Bags - 1 Quart, 1000/cs Superior strength, puncture-resistant poly with heavy-duty zipper for security. 100% virgin polyethylene film. Meets FDA and USDA specifications. Latex free,BHT free (Butylated Hydroxy Toluene), Chlorine free, PVC free. TEMPERATURE: 20° - 150° F