Learn More
Elkay Plastics Company Bag Specimen Transport Seal N Rip Style 3 wall Red Tint with Biohazard STAT print and Outside Pouch, Adhesive closure 1.8 mil 6 x 10 1,000 per Case
Catalog No. NC2743954
Encompass_Preferred
Lab-Seal Tamper-Evident Specimen Bags provide an excellent means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs. The bags have a special mechanism where once its marked tab is pulled, the bag will seal securely and permanently with an adhesive glue. Once the bag is sealed, the only way to access the contents would be to tear off the top indicated by a tear line, which would reveal any tampering.
Catalog No. NC2743954 Supplier Elkay Plastics Company Supplier No. LABA610RST
Lab-Seal Tamper-Evident Specimen Bags provide an excellent means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs. The bags have a special mechanism where once its marked tab is pulled, the bag will seal securely and permanently with an adhesive glue. Once the bag is sealed, the only way to access the contents would be to tear off the top indicated by a tear line, which would reveal any tampering.