Learn More
Elkay Plastics Company Bag Specimen Transport Seal N Rip Style 3 wall Clear with Biohazard print and Outside Pouch, Adhesive Closure 1.8 mil 15 X 17 1,000 per Case
Catalog No. NC2743948
Encompass_Preferred
Lab-Seal Tamper-Evident Specimen Bags provide an excellent means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs. The bags have a special mechanism where once its marked tab is pulled, the bag will seal securely and permanently with an adhesive glue. Once the bag is sealed, the only way to access the contents would be to tear off the top indicated by a tear line, which would reveal any tampering.
Catalog No. NC2743948 Supplier Elkay Plastics Company Supplier No. LABA1517B
Lab-Seal Tamper-Evident Specimen Bags provide an excellent means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs. The bags have a special mechanism where once its marked tab is pulled, the bag will seal securely and permanently with an adhesive glue. Once the bag is sealed, the only way to access the contents would be to tear off the top indicated by a tear line, which would reveal any tampering.