Learn More
Elkay Plastics Company Bag Reclosable Amber UV Protected 3 mil 6 X 8 1,000 per Case
Catalog No. 501049703
Encompass_Preferred
Amber bags prevent their contents from being broken down or damaged by UV light waves. The convenient Zipper closure of these bags make them excellent for packaging and transporting medications and other UV light-sensitive products.
Catalog No. 50-104-9703 Supplier Elkay Plastics Company Supplier No. FAM30608
Amber bags prevent their contents from being broken down or damaged by UV light waves. The convenient Zipper closure of these bags make them excellent for packaging and transporting medications and other UV light-sensitive products.