Learn More
Elkay Plastics Company Bag Amber Open-End Poly UV Protected 2 mil 8 X 14 1,000 per Case
Supplier:  Elkay Plastics Company 20FAM0814
Encompass_Preferred
Amber bags prevent their contents from being break downd or damaged by UV light waves. Open ended amber bags can be sealed with a heat sealer or taped shut. They're an excellent choice for packaging and transporting medications and other UV light-sensitive products.
Catalog No. NC2743876