Learn More
Elkay Plastics Company Bag Amber Open-End Poly UV Protected 2 mil 3 X 9 1,000 per Case
Catalog No. NC2743868
Encompass_Preferred
Amber bags prevent their contents from being break downd or damaged by UV light waves. Open ended amber bags can be sealed with a heat sealer or taped shut. They're an excellent choice for packaging and transporting medications and other UV light-sensitive products.
Catalog No. NC2743868 Supplier Elkay Plastics Company Supplier No. 20FAM0309
Amber bags prevent their contents from being break downd or damaged by UV light waves. Open ended amber bags can be sealed with a heat sealer or taped shut. They're an excellent choice for packaging and transporting medications and other UV light-sensitive products.