Learn More
Chemglass Life Sciences BowTie Biohazard Autoclave Bag, with Drawstring, Large Marking Area and Sterilization Indicator, 25 x 35"
SDP
Catalog No. 50143692
Encompass_Preferred
Autoclave Bag, 25 x 35". Autoclave bags offer maximum protection and performance for autoclavability. The durable 0.002in thick transparent polypropylene bag can be filled with used labware such as Petri dishes, membrane filters, culture plates, pipettes, etc., then autoclaved and discarded. These three large size bags hold 65, 200 and 240 standard size Petri dishes respectively. Steam autoclavable at 135¿C (275¿F). Sold 100 per case.
Catalog No. 50-143-692 Supplier Chemglass Life Sciences Supplier No. CLS11922436
Autoclave Bag, 25 x 35". Autoclave bags offer maximum protection and performance for autoclavability. The durable 0.002in thick transparent polypropylene bag can be filled with used labware such as Petri dishes, membrane filters, culture plates, pipettes, etc., then autoclaved and discarded. These three large size bags hold 65, 200 and 240 standard size Petri dishes respectively. Steam autoclavable at 135¿C (275¿F). Sold 100 per case.