Learn More
Elkay Plastics Company Bag Specimen Transport 3 wall Clear with Removable Biohazard logo, Absorbent Pad and Outside Pouch, Reclosable Closure 1.75 mil 12 X 15 1,000 per Case
Catalog No. NC2743958
Encompass_Preferred
LKs Lab-Loc Reclosable Zipper Specimen Bags with Seal-N-Rip removable biohazard symbols provide an excellent means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs. The bags have a special removable biohazard symbol which can be torn off and allows the bags to be disposed of with regular waste if they are uncontaminated. Absorbent pad in bag helps contain specimen spillage.
Catalog No. NC2743958 Supplier Elkay Plastics Company Supplier No. LABZ1215BA
LKs Lab-Loc Reclosable Zipper Specimen Bags with Seal-N-Rip removable biohazard symbols provide an excellent means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs. The bags have a special removable biohazard symbol which can be torn off and allows the bags to be disposed of with regular waste if they are uncontaminated. Absorbent pad in bag helps contain specimen spillage.