Learn More
Hopkins Medical Products BAG BIO TRANSPORT INSULATED
SDP
Catalog No. NC9744448
Encompass_Preferred
Insulated Bio Transport Bag with outside sleeve and nylon handle. Double stitched in order to be leakproof. Hook and Loop closure on flap. Red with biohazard label.
Catalog No. NC9744448 Supplier Hopkins Medical Products Supplier No. 539656
Insulated Bio Transport Bag with outside sleeve and nylon handle. Double stitched in order to be leakproof. Hook and Loop closure on flap. Red with biohazard label.