Learn More
Chemglass Life Sciences Bag, Cell Culture, Motion (Rocker), 10 L, Ultra Clear, 6-Port (CellGrow), Bioprocess, Single Use
SDP
Catalog No. 5000083157
Encompass
Cell culture bags are manufactured from ultra clear Renolit 9101 film and gamma irradiated. The film meets USP Class VI specifications and is contamination free. The inner product contact material is low density polyethylene. All bags and contact components content materials are USP Class VI tested. Excellent choice for mammalian and plant culture cell lines with extended mixing times.
Catalog No. 50-000-83157 Supplier Chemglass Life Sciences Supplier No. CLS120710
Cell culture bags are manufactured from ultra clear Renolit 9101 film and gamma irradiated. The film meets USP Class VI specifications and is contamination free. The inner product contact material is low density polyethylene. All bags and contact components content materials are USP Class VI tested. Excellent choice for mammalian and plant culture cell lines with extended mixing times.