Learn More
Chemglass Life Sciences Chemglass Bag, Cell Culture, Motion, 20 L, Ultra Clear, 6-Port (Pack of 10), Bioprocess, Single Use
SDP
Catalog No. 502733328
Encompass
Cell culture bags are manufactured from ultra clear Renolit 9101 film and gamma irradiated. The film meets USP Class VI specifications and is contamination free. The inner product contact material is low density polyethylene. All bags and contact components content materials are USP Class VI tested. Excellent choice for mammalian and plant culture cell lines with extended mixing times.
Catalog No. 50-273-3328 Supplier Chemglass Life Sciences Supplier No. CLS120720M
Cell culture bags are manufactured from ultra clear Renolit 9101 film and gamma irradiated. The film meets USP Class VI specifications and is contamination free. The inner product contact material is low density polyethylene. All bags and contact components content materials are USP Class VI tested. Excellent choice for mammalian and plant culture cell lines with extended mixing times.