Learn More
Chemglass Life Sciences Chemglass cell culture bag, 22 L, 6-port, ultra clear (CLS-1207-22)
SDP
Catalog No. 5000083158
Encompass
Chemglass Life Sciences 22 L ultra-clear, six-port cell culture bag designed for rocker and wave bioreactor applications. This single-use bag is intended for mammalian and plant cell cultures, offering visual clarity, sterility, and compatibility with standard rocker trays.
  • 22 L single-use culture bag.
  • Six standard ports for fluid transfer and sampling.
  • Ultra-clear, crack-resistant film for visual inspection.
  • USP Class VI tested product-contact materials.
  • Gamma irradiated for sterility.
  • Compatible with rocker/wave bioreactors and GE Wave platforms.
  • Available as single bag or packs of ten.
Catalog No. 50-000-83158 Supplier Chemglass Life Sciences Supplier No. CLS120722
Add to Cart
Add to Cart
Chemglass Life Sciences 22 L ultra-clear, six-port cell culture bag designed for rocker and wave bioreactor applications. This single-use bag is intended for mammalian and plant cell cultures, offering visual clarity, sterility, and compatibility with standard rocker trays.
  • 22 L single-use culture bag.
  • Six standard ports for fluid transfer and sampling.
  • Ultra-clear, crack-resistant film for visual inspection.
  • USP Class VI tested product-contact materials.
  • Gamma irradiated for sterility.
  • Compatible with rocker/wave bioreactors and GE Wave platforms.
  • Available as single bag or packs of ten.