Learn More
Chemglass Life Sciences Autoclave Bag Holder, for use with CLS-1192-1224; 12" x 24" Bag
SDP
Catalog No. 50143469
Encompass
These epoxy-coated, wire-framed bag holders keep biohazard bags open and ready for use. Just place a biohazard bag into the sturdy wire holder and stretch the bag over the top ring to secure it in place. A separate hooked ring rests over the top to keep the bag in place. Circular shape keeps the bag wide open for easy access. Made from bright, biohazard orange colored epoxy-coated wire. Sold 1 per case. *Bag Not Included
Catalog No. 50-143-469 Supplier Chemglass Life Sciences Supplier No. CLS11931224
These epoxy-coated, wire-framed bag holders keep biohazard bags open and ready for use. Just place a biohazard bag into the sturdy wire holder and stretch the bag over the top ring to secure it in place. A separate hooked ring rests over the top to keep the bag in place. Circular shape keeps the bag wide open for easy access. Made from bright, biohazard orange colored epoxy-coated wire. Sold 1 per case. *Bag Not Included