Learn More
Elkay Plastics Company Bag Hazardous Drug Print with Seal Top Closure 4 mil 12 X 15 1,000 per Case
Catalog No. NC2743887
Encompass_Preferred
Hazardous Drug bags help pharmacies and healthcare providers comply with USP 800 directives. Designed to protect personnel who handle drugs classified by the National Institute for Occupational Safety and Health as hazardous, these bags are used for the transportation and clear identification of hazardous drugs from the pharmacy to the site where they are used.
Catalog No. NC2743887 Supplier Elkay Plastics Company Supplier No. F41215HDB
Hazardous Drug bags help pharmacies and healthcare providers comply with USP 800 directives. Designed to protect personnel who handle drugs classified by the National Institute for Occupational Safety and Health as hazardous, these bags are used for the transportation and clear identification of hazardous drugs from the pharmacy to the site where they are used.