Learn More
Elkay Plastics Company Bag Metalocene Ice 8 pound capacity 1.75 mil 11 X 20 1,000 per Case
Catalog No. NC2743891
Encompass_Preferred
LK Metallocene Ice Bags are made from a production process that yields a special balance of properties that are ideal for ice bag and cold storage applications. These bags have high impact strength, low haze/high gloss and clarity and excellent durability.
Catalog No. NC2743891 Supplier Elkay Plastics Company Supplier No. H20MET
LK Metallocene Ice Bags are made from a production process that yields a special balance of properties that are ideal for ice bag and cold storage applications. These bags have high impact strength, low haze/high gloss and clarity and excellent durability.