Learn More
Elkay Plastics Company Bag Open-Ended Infuser Syringe Flat Poly 1.5 mil 5 X 7 2,000 per Case
Catalog No. NC1910935
Encompass_Preferred
Low Density Polyethylene (LDPE) bags are the most commonly used plastic bag because they provide excellent protection and excellent value. LKPolyethylene bags are made from virgin materials, so they comply with FDA requirements for use in food applications.
Catalog No. NC1910935 Supplier Elkay Plastics Company Supplier No. 15F0507
Low Density Polyethylene (LDPE) bags are the most commonly used plastic bag because they provide excellent protection and excellent value. LKPolyethylene bags are made from virgin materials, so they comply with FDA requirements for use in food applications.