Learn More
Andwin Scientific BAG:POLY;3X5IN;ZIPLOCK 1000PK
SDP
Catalog No. NC9615455
Encompass_Preferred
The Ziploc bags offer secure storage with its zip seal. The bags measure 3" x 5" with a thickness of 2 mil. This product is sold in a pack of 1000.
Catalog No. NC9615455 Supplier Andwin Scientific Supplier No. 90015KY
The Ziploc bags offer secure storage with its zip seal. The bags measure 3" x 5" with a thickness of 2 mil. This product is sold in a pack of 1000.